missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Double Pore Combination pH Electrode
Brand: Hamilton™ 238400
This item is not returnable.
View return policy
Specifications
0 ± 20mV | |
12 mm | |
For use with solid and semi-solid samples | |
0 to 14 |
S7 Electrical Connector | |
Double Pore Combination pH Electrode | |
35 mm | |
0°C to 60°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction