Learn More
Gilac™ Double-wall Container with Stainless Steel Wheel Housing
Double-wall Container with Stainless Steel Wheel Housing is ideal for transporting crushed ice, prepared food and marinated items. Wide range of uses.
Brand:  Gilac™ G157921
 View more versions of this product
Product Code. 17938821
  • 19970.00 SEK / Each
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Ideal for transporting crushed ice, prepared food and marinated items.
  • Wide range of uses.
  • No gaps nor overlaps and no sharp corners.
  • Double-walled offering excellent thermal insulation and excellent mechanical resistance.
  • Container comprising a drainage outlet with drain plug.
  • Rotational molded container with 160 mm, polypropylene wheels with synthetic rubber tires (2 fixed and 2 pivoting) and stainless steel wheel housing.
  • Stainless steel clevis recommended in humid environments (no damage to the bearings even in the presence of water).
  • Can be stored on 800 x 1200 pallets, stackable upside down.
  • Integral high-flow drainage.
  • 1/4 turn angle stop valve optional extra.
  • Volume in litres is equivalent to the container filled to the brim.
Specifications
Specifications
Storage Tote
White
132.1 gal.
HDPE
Rectangular
500 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gilac™ Double-wall Container with Stainless Steel Wheel Housing > Double-wall Container Stainless Steel Wheel Housing; Capacity: 500 L