Learn More
Lovibond™ DPD No. 3 Evo Tablets
This tablet is used in combination with the DPD No. 1 Tablet to measure Free and Total Chlorine concentrations in a water sample.
Brand: Lovibond™ 511420BT
Additional Details : Weight : 0.02000kg
Description
- Shelf-stable, analytical solution for testing water quality parameters with tablet reagents
- Each tablet is protected from humidity, heat and other environmental interferences
- Pharmaceutical-grade aluminum blister packaging individually seals up to 10 tablets on an individual strip
- Easy to dispense blister packaging provides safe handling of the reagents
- Accurate and precise dosing
- Long shelf life
Specifications
Chlorine | |
Water Quality Analysis | |
100 Tests | |
DPD |
Tablet | |
Lovibond™ Colorimeters and Spectrophotometers | |
10 mL | |
2 min |