Learn More
Drabkin's Hemoglobin Reagent For Diagnostics, Chem Lab
Brand: Chem Lab CL02.0401.1000
Specifications
Yellow reagent for diagnostic testing | |
Colored polyethylene | |
1 L | |
Store at room temperature |
Safety and Handling
- Drabkin's Haemoglobine reagent
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P391-Collect spillage.
Supplemental information
- MIXTURE LIST-Contains : potassium cyanide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.