Learn More
Invitrogen™ DRAQ7™ Dye
DRAQ7 Dye is a bright, easy-to-use fluorescent DNA dye used to exclude non-viable cells by flow cytometry.
Brand: Invitrogen™ D15106
Additional Details : Weight : 0.00100kg
Description
This dye has excitation maxima at 599/644 nm and emission maxima at 678/697 nm.
• Stable at room temperature
• Rapid staining of dead cells without wash steps
• No UV excitation and no emission spectral overlap with PE
DRAQ7 Dye is a membrane-impermeable dye that rapidly stains the double-stranded DNA (dsDNA) of dead or permeabilized cells. The dye can be excited by wavelengths from 488 to 647 nm, though it is optimally excited by red laser lines. Its emission maxima is at 697 nm when intercalated with dsDNA.
Specifications
Store at room temperature. | |
Dye | |
200 Assays |
For Research Use Only. Not for use in diagnostic procedures.