Learn More
Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone
Excellent for protecting reactions against moisture while equalizing pressure. Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 05337004
Additional Details : Weight : 0.05400kg
Description
- Tube bent at a 75° angle
- Available in a range of cone sizes
- Corrosion-free and resistant to thermal shock
Specifications
Bent | |
Drying Tube | |
NS 29/32 |