Learn More
Lenz Laborglasinstrumente™ Erlenmeyer Flasks With Conical Ground Joint
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03021937
 View more versions of this product
Product Code. 10298070
This item is not returnable. View return policy
Description
Description
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
DIN EN ISO 4797
Borosilicate Glass, Duran Glass
NS19/26
100 mL
With scale (approx. volume/mL), with inscription label
64 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasinstrumente™ Erlenmeyer Flasks With Conical Ground Joint > Capacity: 100mL; Frosted: NS19/26