Learn More
DWK Life Sciences DURAN™ Funnel, with short stem
A funnel enables a clean transfer and filtration of liquids.
885.00 SEK - 4725.00 SEK
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Autoclavable | Yes |
For Use With (Application) | Decanting and filtration of liquids; The spillage of substances is avoided; Filtering |
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
11772352
|
DWK Life Sciences
213513304 |
55 mm |
2285.00 SEK
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
11782352
|
DWK Life Sciences
213514103 |
80 mm |
2325.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11792352
|
DWK Life Sciences
213514609 |
100 mm |
2600.00 SEK
Pack of 10 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
11702362
|
DWK Life Sciences
213515708 |
150 mm |
4725.00 SEK
Pack of 10 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
10455651
|
DWK Life Sciences
213516104 |
200 mm |
1130.00 SEK
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
10323721
|
DWK Life Sciences
213516901 |
300 mm |
2190.00 SEK
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
11736153
|
DWK Life Sciences
213512805 |
45 mm |
1555.00 SEK
Pack of 10 |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
11746153
|
DWK Life Sciences
213513801 |
70 mm |
2255.00 SEK
Pack of 10 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
13197164
|
DWK Life Sciences
213512308 |
35 mm |
1500.00 SEK
Pack of 10 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
11756153
|
DWK Life Sciences
213515108 |
120 mm |
3235.00 SEK
Pack of 10 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
10758551
|
DWK Life Sciences
213515905 |
180 mm |
885.00 SEK
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
Description
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
- With short stem
- Due to its shape, it is suitable for liquid-solid filtration using a filter paper
- Fired-on, highly durable white print
Specifications
Borosilicate Glass (3.3) | |
Yes | |
Decanting and filtration of liquids; The spillage of substances is avoided; Filtering |