Learn More
DWK Life Sciences DURAN™ Glass Stopper, Clear, Obliquely Chamfered, For oxygen bottles according to Winkler
DURAN™ glass ground stopper according to Winkler
Brand: DWK Life Sciences 246220602
Additional Details : Weight : 0.17000kg
Description
Solid, obliquely chamfered DURAN™ glass stoppers according to Winkler. Stopper for use with oxygen bottles according to Winkler (determination of oxygen dissolved in water) Obliquely cut base prevents air being trapped in the bottle.
- Ground conical, for reagent bottles and oxygen bottles according to Winkler
- All DURAN™ glass components and therefore suitable for storage of aggressive materials
- Tight closure due to standard ground joint
- Octagonal shaped head
Specifications
For use with oxygen bottles according to Winkler (determination of oxygen dissolved in water) | |
10/Pk. | |
Borosilicate Glass | |
Not Autoclavable |