Learn More
DWK Life Sciences DURAN™ Separating Funnel Stopcock with short inlet, complete with PTFE key
For easier attachment to DURAN™ separating funnel raw bodies.
Brand:  DWK Life Sciences 286433007
 View more versions of this product
Product Code. 15894748
  • 17185.00 SEK / Pack of 25
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
DURAN™ Separating funnel taps with short funnel are components for the production of Gilson separating funnels with a particularly strong approach to reducing the risk of breakage with a conical shape right down to the socket of the valve. In the polished version with PTFE plug suitable for grease free working.
  • Separating funnel tap sleeve with PTFE plug
  • with short approach funnel
  • With a particularly strong approach to reducing the risk of breakage
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 mm
Autoclavable
25/Pk.
Borosilicate Glass (3.3), PTFE
18.8 NS
For attachment to DURAN™ separating funnel raw bodies