Learn More
DWK Life Sciences DURAN™ Separating Funnel Stopcock with short inlet, complete with PTFE key
For easier attachment to DURAN™ separating funnel raw bodies.
Brand:  DWK Life Sciences 286432602
 View more versions of this product
Product Code. 15874748
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
DURAN™ Separating funnel taps with short funnel are components for the production of Gilson separating funnels with a particularly strong approach to reducing the risk of breakage with a conical shape right down to the socket of the valve. In the polished version with PTFE plug suitable for grease free working.
  • Separating funnel tap sleeve with PTFE plug
  • with short approach funnel
  • With a particularly strong approach to reducing the risk of breakage
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 mm
Autoclavable
25/Pk.
Borosilicate Glass (3.3), PTFE
12.5 NS
For attachment to DURAN™ separating funnel raw bodies