Learn More
DWK Life Sciences DURAN™ Separating Funnel Stopcock with short inlet, complete with PTFE key
For easier attachment to DURAN™ separating funnel raw bodies.
Brand: DWK Life Sciences 286432808
This item is not returnable.
View return policy
Description
DURAN™ Separating funnel taps with short funnel are components for the production of Gilson separating funnels with a particularly strong approach to reducing the risk of breakage with a conical shape right down to the socket of the valve. In the polished version with PTFE plug suitable for grease free working.
- Separating funnel tap sleeve with PTFE plug
- with short approach funnel
- With a particularly strong approach to reducing the risk of breakage
- Glass articles for further processing
Specifications
2.5 mm | |
Autoclavable | |
25/Pk. |
Borosilicate Glass (3.3), PTFE | |
14.5 NS | |
For attachment to DURAN™ separating funnel raw bodies |