Learn More
DWK Life Sciences DURAN™ Straight Burette Valve, GU
Through burette valve as an economical alternative.
1095.00 SEK
Specifications
Material | Borosilicate Glass (3.3), PTFE |
---|---|
Autoclavable | Autoclavable |
No. per Pack | 10/Pk. |
For Use With (Application) | GU straight through burette valve valves are suitable for lower leakage rate requirements, e.g. for flow control of liquids; Ideal for dosing and controlling liquids |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11600049
|
DWK Life Sciences
286059208 |
For Burette 50 mL | N/A | ||||||
Description
Valves GU have an unscrewing safety device which prevents the valve pin from being unscrewed absolutely. The adjustable valve pin increases the service life of the valve tap. GU Valves are suitable for lower leakage rate requirements, e.g. for flow control of liquids or gases.
- Through burette valve valve as low-cost alternative
- Media flowing through the valve only come into contact with DURAN™ glass and PTFE.
- Grease-free working
- Handy design of the valve allows easy disassembly
- The unscrewing protection prevents absolute unscrewing of the valve pin.
- Valve pin adjustable to increase the service life of the valve tap
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3), PTFE | |
Autoclavable | |
10/Pk. | |
GU straight through burette valve valves are suitable for lower leakage rate requirements, e.g. for flow control of liquids; Ideal for dosing and controlling liquids |