missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP22 Mouse anti-Human, Mouse, Rabbit, Rat, Clone: 3B8F1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68032-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
DUSP22 Monoclonal antibody specifically detects DUSP22 in Human, Mouse, Rabbit, Rat samples. It is validated for IF, WB, ELISASpecifications
DUSP22 | |
Monoclonal | |
1000 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
Mouse | |
20 μL | |
56940 | |
Store at -20°C | |
Liquid |
Immunofluorescence, Western Blot, ELISA | |
3B8F1 | |
Unconjugated | |
BC006459 | |
Protein A purification | |
Primary | |
Human, Mouse, Rabbit, Rat | |
Antibody | |
IgG2a |