missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP22 Mouse anti-Human, Mouse, Rabbit, Rat, Clone: 3B8F1, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | DUSP22 |
---|---|
Clone | 3B8F1 |
Concentration | 1000 μg/mL |
Applications | Immunofluorescence, Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17741139
|
Proteintech
68032-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17731139
|
Proteintech
68032-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DUSP22 Monoclonal antibody specifically detects DUSP22 in Human, Mouse, Rabbit, Rat samples. It is validated for IF, WB, ELISASpecifications
DUSP22 | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
Human, Mouse, Rabbit, Rat | |
56940 | |
Primary | |
Store at -20°C |
3B8F1 | |
Immunofluorescence, Western Blot, ELISA | |
Unconjugated | |
Mouse | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
IgG2a | |
Protein A purification | |
Antibody |