missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynactin 4 Monoclonal Antibody (3G9D7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517065
This item is not returnable.
View return policy
Description
MA5-17065 targets DCTN4 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human samples. The MA5-17065 immunogen is purified recombinant fragment of human DCTN4 (amino acids: 57-298) expressed in E. Coli. MA5-17065 detects DCTN4 which has a predicted molecular weight of approximately 52.3kDa.
May play an role in the regulation of Ins(1,4,5)P3 around the endoplasmic reticulum. Target protein's p62 subunit of the dynactin complex involved in the cytoplasmic dynein motor machinery.Specifications
Dynactin 4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9UJW0 | |
DCTN4 | |
Purified recombinant fragment of human DCTN4 (amino acids: 57-298) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
3G9D7 | |
Unconjugated | |
DCTN4 | |
1110001K06Rik; 4930547K17Rik; C130039E17Rik; DCTN4; Dyn4; dynactin 4; dynactin 4 (p62); dynactin 4 (p62) isoform 1; dynactin 4 (p62) isoform 2; dynactin p62 subunit; dynactin subunit 4; Dynactin subunit p62; hypothetical protein LOC550479; P62; si:ch211-174h24.2; zgc:112431 | |
Mouse | |
Protein G | |
RUO | |
51164 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |