Learn More
Abeba™ Dynamic 7313 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E, FO SRC |
---|---|
Color | White |
Material | Leather |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15692658
|
Abeba™
7313/35 |
35 |
N/A
|
N/A | |||||
15602668
|
Abeba™
7313/36 |
36 |
N/A
|
N/A | |||||
15612668
|
Abeba™
7313/37 |
37 |
N/A
|
N/A | |||||
15622668
|
Abeba™
7313/38 |
38 |
N/A
|
N/A | |||||
15632668
|
Abeba™
7313/39 |
39 |
N/A
|
N/A | |||||
15642668
|
Abeba™
7313/40 |
40 |
N/A
|
N/A | |||||
15652668
|
Abeba™
7313/41 |
41 |
N/A
|
N/A | |||||
15662668
|
Abeba™
7313/42 |
42 |
N/A
|
N/A | |||||
15672668
|
Abeba™
7313/43 |
43 |
N/A
|
N/A | |||||
15682668
|
Abeba™
7313/44 |
44 |
N/A
|
N/A | |||||
15692668
|
Abeba™
7313/45 |
45 |
N/A
|
N/A | |||||
15602678
|
Abeba™
7313/46 |
46 |
N/A
|
N/A | |||||
15612678
|
Abeba™
7313/47 |
47 |
N/A
|
N/A | |||||
15622678
|
Abeba™
7313/48 |
48 |
N/A
|
N/A | |||||
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather white/red
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Polyurethane |
White | |
Slip-resistant | |
7313 |