Learn More
Tocris Bioscience™ E 64
Potent and irreversible cysteine protease inhibitor
Brand:  Tocris Bioscience™ 5208/50
 View more versions of this product
Product Code. 18790288
  • 9665.00 SEK / 50mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
E 64 is a potent and irreversible cysteine protease inhibitor (IC50 values are 1.4, 2.5 and 4.1 nM for cathepsin K, L and S respectively. Also inhibits papain, calpain, cathepsin B and cathepsin H. Has no effect on serine proteases. Reduces HSC-3 tongue cancer cell invasion and induces oxidative stress and apoptosis in filarial parasites. Also inhibits autophagy-associated lysosomal degradation in vitro.
Specifications
Specifications
>98%
E 64
E 64
50 mg
357.41