Learn More
3M™ E-A-Rfit™ Dual-Ear Validation System
Click to view available options
Hearing Protector Fit Test Kit 393-1100
Description
- Objective, dual-ear test, takes only seconds to administer for seven standard frequencies from 125Hz to 8000Hz
- Includes software, speaker and impact-resistant carrying case
- Free online training and user support
- Requires probed test plugs, pods, or cushions
- Dual-ear design saves time by measuring both ears simultaneously in under five seconds
- Dual-element, dual microphone assembly is the key to the system, measuring sound levels inside and outside the hearing protector
- Fast, clear, accurate results are obtained in just seconds and can be printed or saved electronically for future reference
- System determines personal attenuation rating (PAR) of individual workers and helps remove measurement, fitting, and spectral uncertainty
- Requires probed test plugs
Specifications
Specifications
Includes | Software, speaker and impact resistant carrying case |
Certifications/Compliance | CE Approved |
For Use With (Application) | For automotive, food processing, general manufacturing, military, maintenance, mining, petroleum, pharmaceutical and transportation sectors |
Quantity | 1 kit |
Product Type | Hearing Protector Fit Test Kit 393-1100 |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ E-A-Rfit™ Dual-Ear Validation System >
Spot an opportunity for improvement?Share a Content Correction