Learn More
STEMCELL Technologies EasySep™ Human EpCAM Positive Selection Kit II
Immunomagnetic positive selection of human EpCAM+ cells (e.g. mammary epithelial cells)
Brand: STEMCELL Technologies 17846
Description
- The EasySep™ Human EpCAM Positive Selection Kit II is designed to isolate EpCAM+ cells from fresh or previously frozen human mammary epithelial cell preparations by immunomagnetic positive selection
- Desired cells are targeted with antibody complexes recognizing EpCAM and dextran-coated magnetic particles
- The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- The EpCAM+ fraction contains luminal cells, luminal-restricted, and bipotential progenitor cells
- This product replaces the EasySep™ Human EpCAM Positive Selection Kit (Catalog #18356)
Specifications
Mammary Cells | |
Cell Isolation | |
1 x 109 cells |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.