Learn More
STEMCELL Technologies EasySep™ Human Plasmacytoid DC Enrichment kit
Immunomagnetic isolation of untouched human plasmacytoid dendritic cells (pDC)
Brand: STEMCELL Technologies 19062
This item is not returnable.
View return policy
Description
- The EasySep™ Human Plasmacytoid DC Enrichment Kit is designed to isolate plasmacytoid dendritic cells (pDC) from fresh peripheral blood mononuclear cells by negative selection
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-pDCs and dextran-coated magnetic particles
- The labeled cells are separated using the EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
Dendritic Cells | |
Cell Isolation | |
2 x 109 cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.