Learn More
STEMCELL Technologies EasySep™ Mouse Naïve CD4+ T Cell Isolation Kit
Immunomagnetic isolation of untouched mouse naive CD4+ T cells
Brand: STEMCELL Technologies 19765
This item is not returnable.
View return policy
Description
- The EasySep™ Mouse Naïve CD4+ T Cell Isolation Kit is designed to isolate naïve CD4+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection
- Unwanted cells are targeted for removal with biotinylated antibodies directed against non-naïve CD4+ T cells (CD8, CD11b, CD11c, CD19, CD24, CD25, CD44, CD45R, CD49b, TCRγ/δ, TER119) and streptavidin-coated magnetic particles (RapidSpheres™ )
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
T Cells, T Cells, CD4+ | |
Cell Isolation | |
1 x 109 cells |