missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ ECO AluRack for Chest Freezers, 100 mm Box Height

ECO AluRack fits into any chest freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE11564

Additional Details : Weight : 1.60000kg

 View more versions of this product

Product Code. 12448150

  • 810.00 SEK / Each
Estimated Shipment: 09-10-2024
to see stock.
Add to basket

Description

Description

  • Made of seawater resistant aluminum
  • Fits into any freezer and can be used with all standard cryoboxes
  • Racks can be used with all freezer brands
  • Without standard white cardboard boxes
Specifications

Specifications

Seawater resistant
Aluminum
Chest Frseezers
100 mm (Box)
Boxes
5
140 x 140 x 530 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers