missing translation for 'onlineSavingsMsg'
Learn More

Medline Scientific™ Economy Standard Incubator, 205 L

Temperature Range: 3°C to 60°C

Brand:  Medline Scientific™ IB-21E

Product Code. 12823486

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The E Series sizes are 65L, 151L and 205L with a temperature range ambient 3°C to 60°C. Film heaters located externally on the chamber sides makes the unit completely vibration free.The G Series sizes are 60L, 102L and 151L with a temperature range ambient 5°C to 70°C. The chamber is air jacketed and uses convection heating. The G Series also comes with Labtracer software to both control and data log the unit via PC.
  • Two series available dependent on temperature range and size required
  • Stainless steel inner chamber with rounded corners
  • Inner glass door for chamber inspection
  • Two shelves supplied as standard
  • Programmable with LED display of set and actual temperatures (0.1°C resolution)
  • Open-door alarm
Bench top heated incubators for use where a stable environment for cell growth or other applications is required.
Specifications

Specifications

205 L
3°C to 60°C
740 mm
515 mm
230 V
2
515 x 632 x 630 mm
632 mm
57 kg
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.