Learn More
Schuett Biotec™ EcoVac Safety Extraction System 4
The EcoVac Safety Extraction System 4 is for safe and convenient aspiration of liquid supernatants in cell culture. Increases work safety when handling infectious material. Autoclavable, unbreakable and resistant to chemicals.
Brand:  Schuett Biotec™ 3350042
Product Code. 10308961
  • 4090.78 SEK / Each
Estimated Shipment: 04-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • For aspiration of liquid supernatants in cell culture
  • Increases work safety
  • Completely autoclavable, unbreakable and resistant to chemicals
  • Quick-release couplings automatically close the bottle and hermetically seal it for transport
Specifications
Specifications
300 mbar
EcoVac system 4, 4 L safety collection bottle with screw cap, 2 quick-release couplings, 2 angle nipples for hose connections, 2 x 1.5 m silicone hose, ventilation insert and safety filter
4 L/min.