missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EED Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534430
This item is not returnable.
View return policy
Description
A suggested positive control is human heart tissue lysate. PA5-34430 can be used with blocking peptide PEP-1472.
The EED protein (WAIT-1 or WD protein associated with integrin cytoplasmic tails-1), also called embryonic ectoderm development, is a member of the superfamily of WD-40 repeat proteins and widely conserved polycomb group (PcG) family of proteins. The polycomb group (PcG) is a large and evolutionarily conserved set of genes whose products act in multimeric complexes to modify histones, which are then thought to cause stable and heritable states of transcriptional repression. EED shuttles between the nucleus and the plasma membrane and can interact with the cytoplasmic tail of integrin Beta-7 subunit. EED exerted an antiviral activity at the late stage of HIV-1 replication.Specifications
EED | |
Polyclonal | |
Unconjugated | |
EED | |
EED; embryonic ectoderm development; Embryonic ectoderm development protein; ENSMUSG00000039373; HEED; l(7)5Rn; l7Rn5; lethal, Chr 7, Rinchik 5; lusk; Polycomb protein EED; WAIT1; WAIT-1; WD protein associating with integrin cytoplasmic tails 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13626, 293104, 8726 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
ChIP Assay, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O75530, Q921E6 | |
EED | |
An 18 amino acid synthetic peptide near the amino terminus of human EED | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |