Learn More
Lonza™ EGM™-Plus Endothelial Cell Growth Media-Plus BulletKit™
Complete kit, growth medium and supplements, to support growth of human umbilical vein endothelial cells.
Brand:  Lonza™ CC-5035
Product Code. 15886064
  • 5720.00 SEK / 1 set
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Culture kit containing growth medium and supplements.
  • Developed to support optimal growth of human umbilical vein endothelial cells.
  • Includes the following:
    • 1 x EBM™ Plus Basal Medium (CC-5036), 475 mL
    • 1 x EGM™Plus SingleQuots™ Supplement Pack (CC-4542) containing the following:
      • 1 x Bottle FBS, 10.00 mL
      • 1 x Vial Hydrocortisone, 0.50 mL
      • 1 x Bottle L-Glutamine, 25.00 mL
      • 1 x Vial EnGS, 1.00 mL
      • 1 x Vial Ascorbic Acid, 0.50 mL
      • 1 x Vial rhEGF, 0.50 mL
      • 1 x Vial GA-1000, 0.50 mL
      • 1 x Vial Heparin, 0.50 mL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Basalmedium, Fetales Kälberserum, Hydrocortison, L-Glutamin, EnGS, Ascorbinsäure, rhEGF, GA-1000, Heparin
Endothelial Cells
Growth Medium and Supplements
Medium: 475 mL, FBS: 10.0 mL, Hydrocortisone: 0.50 mL, L-Glutamine: 25.0 mL, EnGS: 1.0 mL, Ascorbic Acid: 0.5 mL, rhEGF: 0.5 mL, GA-1000: 0.5 mL, Heparin: 0.50 mL