Learn More
Bohlender™ Elbow Shaped PTFE Tube Fittings
Distribute liquids or gases with this tube fitting. BOLA GL Elbow-shaped Tube Fittings serve as a reduction for connecting different diameters of tubes or tubing.
Brand:  Bohlender™ D858-18
Product Code. 10584145
  • 514.00 SEK / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Elbow-shaped tube fitting made of PTFE-EX
  • Two connections with GL thread
  • For connection of tube with BOLA Laboratory Screw Joints