missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566521
This item is not returnable.
View return policy
Description
Immunogen sequence: QDCHSHSRTSL QSSHLWEFVR DLLLSPEENC GILEWEDREQ GIFRVVKSEA LAKMW Highest antigen sequence identity to the following orthologs - mouse 96%, rat 100%.
ELF5 is a member of an epithelium-specific subclass of the Ets transcritpion factor family. In addition to its role in regulating the later stages of terminal differentiation of keratinocytes, it appears to regulate a number of epithelium-specific genes found in tissues containing glandular epithelium such as salivary gland and prostate. It has very low affinity to DNA due to its negative regulatory domain at the amino terminus.Specifications
ELF5 | |
Polyclonal | |
Unconjugated | |
ELF5 | |
E74 like ETS transcription factor 5; E74-like factor 5; E74-like factor 5 (ets domain transcription factor); E74-like factor 5 ESE-2a; Elf5; elf-5; Epithelium specific Ets-2; epithelium-restricted ESE-1-related Ets factor; Epithelium-specific Ets transcription factor 2; ESE2; ESE-2; ESE-5; ESE-5.; ets transcription factor; ETS-related transcription factor Elf-5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2001 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKW6 | |
ELF5 | |
Recombinant Human ELF5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only