Learn More
BD ELISPOT Sets
Sufficient capture antibody, detection antibody, and streptavidin-horseradish peroxidase for 10 x 96 wells
Brand:  BD 551085
 View more versions of this product
Product Code. 15846518
  • 15610.00 SEK / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Includes: Unlabeled capture antibody (no azide/low endotoxin format), Biotinylated detection antibody, Streptavidin-HRP, 10 ELISPOT plates, Certificate of Analysis providing lot-specific optimal reagent concentrations
This item is not returnable. View return policy
Description
Description
  • High sensitivity
  • High throughput, high content analysis
  • Minimal volume of biological material required
  • Suitable for frozen/thawed biological samples
  • Compatibility with other assays—cells analyzed by BD ELISPOT Sets can be used for cloning, proliferation assays, flow cytometry, or other methods of analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-5
Unlabeled capture Antibody, detection Antibody, Streptavidin HRP, 10 ELISPOT plates
4°C
Human IL-5 ELISPOT set
Human
10 x 96 Tests
Cloning, proliferation assays, flow cytometry or other methods of analysis