missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ellis van Creveld syndrome Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1935.00 SEK - 4555.00 SEK
Specifications
Antigen | Ellis van Creveld syndrome |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18332524
|
Bio-Techne
NBP3-03796-20UL |
20 μL |
1935.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18370987
|
Bio-Techne
NBP3-03796-100UL |
100 μL |
4555.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ellis van Creveld syndrome Polyclonal specifically detects Ellis van Creveld syndrome in Mouse samples. It is validated for Western Blot.Specifications
Ellis van Creveld syndrome | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
2121 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
Ellis van Creveld syndrome, ellis-van Creveld syndrome protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 743-992 of human Ellis van Creveld syndrome (NP_714928.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title