Learn More
Corning™ Elplasia™ Microcavity Plates
The only plates with a simple 'plug and play' protocol for scaffold-free, self-assembly spheroid formation at large volumes.
Brand: Corning™ 4444
Additional Details : Weight : 4.17230kg
Description
- Novel well geometry and surface coatings, including Corning Ultra Low Attachment (ULA) featured on the round bottom plates
- Corning ULA surface is hydrophilic, biologically inert and non-degradable, which promotes highly reproducible spheroid formation and easy harvesting
- Corning Elplasia round bottom plates optimal for bulk spheroid formation, collection and expansion\
- Round bottom plates feature Corning Ultra-Low Attachment surface and are available in 6-, 24- and 96-well formats
- Corning Elplasia square bottom plates feature a surface with optical qualities suited for image analysis, making them an ideal solution for clonal selection and high magnification imaging of very small clusters
- The Elplasia square plate is well suited for screening programs as it generates a high volume of spheroids per well, allowing for higher signals per well
- Square bottom plates are plasma-treated for self-coating and come in 6-, 24-, 96- and 384-well formats
- Spheroids may be formed and cultured for 21 or more days
- Can produce between 79 – 15,000+ spheroids per well, depending on plate format, under one culture condition
- The high volume of spheroids generated in each well increase signals per well without increasing spheroid size
- Black opaque sidewalls to reduce well-to-well 'cross-talk'
- Well suited for fluorescent/luminescent assays
- Available in multiple formats, two well geometries and surface coating options
Specifications
Clear | |
Polystyrene | |
Round | |
Polystyrene | |
Sterile | |
Microcavity Plate | |
Lid |
Closure Included | |
Not Autoclavable | |
Black, Clear | |
Rectangular | |
Tissue Culture | |
6 | |
5 |