Learn More
Merck Color Standards Chart
Filter color standards for fluid sampling kits
730.00 SEK - 2350.00 SEK
Product Code | Brand | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||
10106730
|
Merck
ASTM03701 |
For sampling turbine fuels, water or other liquids through Millipore contamination analysis monitors prior to gravimetric or colorimetric analysis. | ASTM Color Chart |
2350.00 SEK
Each |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
10134263
|
Merck
XX6210000 |
Membrane filter comparison | Babcock and Wilcox Color Chart |
730.00 SEK
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
Description
- ASTM03701 is an ASTM color standards chart
- XX6210000 is a Babcock and Wilcox color standards chart
Specifications
Science |