missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emerin (Papillary Thyroid Carcinoma and EDMD Marker) Mouse, Clone: EMD/2167, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2010-MSM1-P0
This item is not returnable.
View return policy
Specifications
Emerin (Papillary Thyroid Carcinoma and EDMD Marker) | |
Nuclear Membrane | |
EMD/2167 | |
Unconjugated | |
Mouse | |
37kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
2010 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human Emerin protein fragment (apprx. aa 56-167) | |
K-562, HeLa or Jurkat cells. Kidney or Skin | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Emerin (Papillary Thyroid Carcinoma and EDMD Marker) Mouse, Clone: EMD/2167, enQuireBio™ > 20 μg
Spot an opportunity for improvement?Share a Content Correction