missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENDOD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA552701
This item is not returnable.
View return policy
Description
Immunogen sequence: GECDKFFYAG TPPAGLAADS HVKICQRAEG AERFATLYST RDRIPVYSAF RAPRPAPGGA EQRWLVEPQI DDPNSNLEEA INEAEAITSV NSLGSKQALN TDYLDSDYQR GQLYPFSLSS DVQVATFTLT NSAPMTQSF Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 21%.
May act as a DNase and a RNase.Specifications
ENDOD1 | |
Polyclonal | |
Unconjugated | |
ENDOD1 | |
2210414F18Rik; 2310067E08Rik; C85344; Endod1; endonuclease domain containing 1; endonuclease domain-containing 1 protein; Kiaa0830; RGD1310444 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23052 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94919 | |
ENDOD1 | |
Recombinant protein corresponding to Human ENDOD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only