Learn More
Endoglin/CD105 Mouse anti-Human, Unconjugated, Clone: 2D5E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67075-1-IG-20UL
This item is not returnable.
View return policy
Description
The Endoglin/CD105 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Endoglin/CD105. This antibody recognizes human antigen. The Endoglin/CD105 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Endoglin/CD105 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA | |
2D5E8 | |
Unconjugated | |
20 μL | |
2022 | |
IgG2b |