Learn More
Lonza™ Endothelial Basal Medium
Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Endothelial Basal Medium is part of a collection of tools optimized to grow certain cell types.
Brand: Lonza™ CC-3121
This item is not returnable.
View return policy
Description
Media products are supplied in several formats:- Basal Medium -– contains no growth factors, cytokines, or supplements
- SingleQuots™ Kit – growth factors, cytokines, and supplements
- BulletKit™ – basal medium and SingleQuots™ Kit packaged together
- EGM™-2 BulletKit™ – Optimized for certain HUVECs, Aortic and Pulmonary artery endothelial cells. Contains 2% FBS and VEGF for rapid proliferation. Does not contain BBE.
- EGM™ BulletKit™ – Optimized for HUVECs cat. no. CC-2517 and CC-2519, contains 2% FBS and BBE. Does not contain exogenous VEGF.
- New! EGM™-Plus BulletKit™ – Enhanced version of EGM™ Medium. Optimized for HUVECs cat. no. CC-2935. Contains 2% FBS and BBE. Does not contain exogenous VEGF. Faster proliferation than HUVECs isolated in EGM™ Medium.
- EGM™-2MV BulletKit™ – Coronary and Iliac artery, human microvascular endothelial cells, contains 5% FBS and VEGF for rapid proliferation. Does not contain BBE.
- EGM™ -MV BulletKit™ – Bovine Microvascular Cells, contains 5% FBS and BBE. Does not contain exogenous VEGF.
Order Info
dimProductType
Specifications
Basal Medium |