Learn More
Lonza™ Endotoxin Challenge Vials™
Features >1,000 EU per vial. Lonza™ Endotoxin Challenge Vials™ are pre-filled vials of endotoxin indicator for depyrogenation in oven validation studies. They can be used with any Lonza endotoxin detection kit.
Brand: Lonza™ 192568
Description
The Endotoxin Challenge Vial™ (ECV) is used in the validation of dry heat depyrogenation cycles. The ability of a particular oven cycle to destroy/inactivate endotoxin is measured by comparing the endotoxin levels in baked ECVs vs unbaked control ECVs. The United States Pharmacopeia (USP) states that the endotoxin indicators used in depyrogenation process challenges allow for accurate indication of at least a 3-log reduction in USP Endotoxin Units.
- Designed to indicate a minimum 3-log reduction in endotoxin content
- Contains > 1,000 EU of purified E. coli 05:B5 endotoxin per vial
- Does not contain any fillers or stabilizers
- For use in oven validation studies
- Can be used with any Lonza endotoxin detection kit:
- PYROGENT™
- QCL-1000™
- Kinetic-QCL™
- PYROGENT™-5000 Limulus Amebocyte Lysate (LAL) Assays
Specifications
USP | |
Lyophilized | |
No Fillers, No Stabilizers | |
Cardboard box with foam insert | |
25 vials |
25 Vials with Stoppers, Store at 2–8°C | |
Endotoxin indicator | |
>1,000 EU/vial | |
Endotoxin detection | |
E. coli 05:B5 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.