Learn More
Brady™ Engraved Replacement Labels for M710, M610, M611
Engraved Plate Replacement Labels for use with Brady™ Printers M611, M610 and M710.
780.00 SEK - 3540.00 SEK
Specifications
Adhesive Type | Permanent Acrylic, Foam-backed |
---|---|
Coating | None |
Finish | Gloss |
Format | Label |
Material | Polyester |
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
17837864
|
Brady™
312072 |
White | 7.95 x 27 mm |
885.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
||||
17807904
|
Brady™
312095 |
White | 12.45 x 27 mm |
1150.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
||||
17808004
|
Brady™
312105 |
White | 15 x 27 mm |
885.00 SEK
Pack of 150 |
In Stock
Log in to see stock. |
||||
17847904
|
Brady™
312113 |
White | 18 x 27 mm |
1010.00 SEK
Pack of 150 |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
17827994
|
Brady™
312120 |
White | 40 x 30 mm |
1090.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||
17847924
|
Brady™
312124 |
White | 75 x 25 mm |
3540.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||
17827924
|
Brady™
312068 |
Black | 7.95 x 27 mm |
885.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
||||
17867894
|
Brady™
312091 |
Black | 12.45 x 27 mm |
1155.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
||||
17857994
|
Brady™
312102 |
Black | 15 x 27 mm |
885.00 SEK
Pack of 150 |
In Stock
Log in to see stock. |
||||
17817924
|
Brady™
312110 |
Black | 18 x 27 mm |
1010.00 SEK
Pack of 150 |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
17807994
|
Brady™
312116 |
Black | 40 x 30 mm |
1090.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||
17817874
|
Brady™
312071 |
Silver | 7.95 x 27 mm |
885.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
||||
17837954
|
Brady™
312094 |
Silver | 12.45 x 27 mm |
1155.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
||||
17867914
|
Brady™
312104 |
Silver | 15 x 27 mm |
885.00 SEK
Pack of 150 |
In Stock
Log in to see stock. |
||||
17817994
|
Brady™
312112 |
Silver | 18 x 27 mm |
1010.00 SEK
Pack of 150 |
In Stock
Log in to see stock. |
||||
17808084
|
Brady™
312119 |
Silver | 40 x 30 mm |
1090.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||
17877874
|
Brady™
312070 |
Red | 7.95 x 27 mm |
780.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
||||
17847984
|
Brady™
312093 |
Red | 12.45 x 27 mm |
1020.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
||||
17887994
|
Brady™
312103 |
Red | 15 x 27 mm |
780.00 SEK
Pack of 150 |
In Stock
Log in to see stock. |
||||
17807854
|
Brady™
312111 |
Red | 18 x 27 mm |
895.00 SEK
Pack of 150 |
In Stock
Log in to see stock. |
||||
17828054
|
Brady™
312118 |
Red | 40 x 30 mm |
1090.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||
17807944
|
Brady™
312073 |
Yellow | 7.95 x 27 mm |
780.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
||||
17867984
|
Brady™
312096 |
Yellow | 12.45 x 27 mm |
1020.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
||||
17897944
|
Brady™
312106 |
Yellow | 15 x 27 mm |
780.00 SEK
Pack of 150 |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
17807914
|
Brady™
312114 |
Yellow | 18 x 27 mm |
895.00 SEK
Pack of 150 |
In Stock
Log in to see stock. |
||||
17847994
|
Brady™
312121 |
Yellow | 40 x 30 mm |
1090.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||
17898034
|
Brady™
312125 |
Yellow | 75 x 25 mm |
3540.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||
17897864
|
Brady™
312069 |
Green | 7.95 x 27 mm |
780.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
||||
17848074
|
Brady™
312092 |
Green | 12.45 x 27 mm |
1020.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
||||
17888034
|
Brady™
312117 |
Green | 40 x 30 mm |
1090.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
- Engraved Plate Replacement labels for the M610 and M611 Thermal Transfer Portable Printers
- Parts can also be printed on the M710 Label Printer provided the roll adapter (M71-ADAPT) is used
- Aggressive adhesive and solvent resistance makes them an ideal replacement for engraved tags
- Service temperature of -40°C to +100°C
Specifications
Permanent Acrylic, Foam-backed | |
Gloss | |
Polyester | |
Thermal Transfer | |
Non-sterile | |
M610, M710, M611, BMP61, BMP71 | |
-40°C | |
-40°C to +100°C | |
0.65 mm |
None | |
Label | |
Single-side Printable | |
Engraved Plate Labels | |
Data and Telecommunications, Electrical Labeling, General Identification | |
1 Label | |
-40°F to +212°F | |
0.0256 in. |