missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENPP5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA553515
This item is not returnable.
View return policy
Description
Immunogen sequence: KFWEEATPIW ITNQRAGHTS GAAMWPGTDV KIHKRFPTHY MPYNESVSFE DRVAKIIEWF TSKEPINLGL LYWEDPDDMG HHLGPDSPLM GPVISDIDKK LGYLIQMLKK AKLWNTLNLI ITSDHGMTQC SE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 86%.
This gene encodes a type-I transmembrane glycoprotein. Studies in rat suggest the encoded protein may play a role in neuronal cell communications. Alternatively spliced transcript variants have been described.Specifications
ENPP5 | |
Polyclonal | |
Unconjugated | |
ENPP5 | |
AW061004; D17Abb1e; EC 3.1; ectonucleotide pyrophosphatase/phosphodiesterase 5; ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative); ectonucleotide pyrophosphatase/phosphodiesterase family member 5; E-NPP 5; ENPP5; E-NPP5; KIAA0879; mKIAA4152; Npp5; NPP-5; UNQ550/PRO1107 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
59084 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UJA9 | |
ENPP5 | |
Recombinant protein corresponding to Human ENPP5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only