Learn More
Sartorius Entris™ II Advanced Analytical Balances
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
34910.00 SEK - 49230.00 SEK
Specifications
Certifications/Compliance | CE Certificate, CSA for Canada |
---|---|
Connection Type | D-sub connector, 9-pin; USB UTL; USB Device |
Display Type | Intuitive Graphic Touch Technology |
Readability | 0.1 mg |
Material | Glass, PBT, Stainless Steel |
Product Code | Brand | Capacity | Calibration | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Calibration | Price | Quantity & Availability | ||||
16605693
![]() |
Sartorius
BCA64I-1S |
60 g | Internal |
37870.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16675683
![]() |
Sartorius
BCA64-1S |
60 g |
34910.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16642454
![]() |
Sartorius
BCA124I-1S |
120 g | Internal |
38815.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16632454
![]() |
Sartorius
BCA124-1S |
120 g |
35975.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16655653
![]() |
Sartorius
BCA224I-1S |
220 g | Internal |
42365.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16625653
![]() |
Sartorius
BCA224-1S |
220 g |
39525.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16646253
![]() |
Sartorius
BCA324I-1S |
320 g | Internal |
49230.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Entris™ II Advanced balances - Quality, Value, and Consistency
- Accurate weighing results that can be trusted time after time
- Simplifies balance leveling with built-in, real-time level support
- Increases reliability of weighing results using integrated protection systems
- Controlled access to balance settings with user management
- Gapless documentation thanks to CalAuditTrail
Specifications
CE Certificate, CSA for Canada | |
Intuitive Graphic Touch Technology | |
Glass, PBT, Stainless Steel | |
≤1.5 sec. | |
100/240 V | |
50/60 Hz |
D-sub connector, 9-pin; USB UTL; USB Device | |
0.1 mg | |
Electronic Analytical Balance | |
Balance | |
0.2 A | |
Class I |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sartorius Entris™ II Advanced Analytical Balances