Learn More
Memmert™ Entry Port 23 mm for UF110plus, Custom Position TBC Left Side
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF110plusF4
Product Code. 17518975
  • 2205.00 SEK / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Description
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in left side
Specifications
Specifications
UF110plus Oven
Entry Port
23 mm
Documents
One moment while we fetch your results.
Product Title
Memmert™ Entry Port 23 mm for UF110plus, Custom Position TBC Left Side > For Use With: UF110plus