Learn More
Memmert™ Entry Port 23 mm for UF110plus, Standard Position Left Side Center/Center
Optional entry port for Memmert Ovens.
Brand: Memmert™ UF110plusF0
This item is not returnable.
View return policy
Description
- Entry port with 23 mm diameter
- Flap and stopper closure
- Standard position left side center/center
Specifications
UF110plus Oven | |
Entry Port | |
23 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Memmert™ Entry Port 23 mm for UF110plus, Standard Position Left Side Center/Center > For Use With: UF110plus
Spot an opportunity for improvement?Share a Content Correction