Learn More
Memmert™ Entry Port 23 mm for UF160plus, Standard Position Left Side Center/Center
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF160plusF0
Product Code. 17569075
  • 1485.00 SEK / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Description
  • Entry port with 23 mm diameter
  • Flap and stopper closure
  • Standard position left side center/center
Specifications
Specifications
UF160plus Oven
Entry Port
23 mm
Product Title
Memmert™ Entry Port 23 mm for UF160plus, Standard Position Left Side Center/Center > For Use With: UF160plus