Learn More
Memmert™ Entry Port 38 mm for UF55plus
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF55plusF7
Product Code. 17559505
  • 2525.00 SEK / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Description
  • Entry port with 38 mm clear diameter
  • Flap and stopper closure
  • Custom position TBC in the back
Specifications
Specifications
UF55plus Oven
Entry Port
38 mm
Documents
One moment while we fetch your results.
Product Title
Memmert™ Entry Port 38 mm for UF55plus > For Use With: UF55plus