Learn More
Memmert™ Entry Port 57 mm for UF160plus
Optional entry port for Memmert Ovens.
Brand: Memmert™ UF160plusF8
This item is not returnable.
View return policy
Description
- Entry port with 57 mm diameter
- Flap closure
- Custom position TBC in the back
Specifications
UF160plus Oven | |
Entry Port | |
57 mm |
Product Content Correction
Product Title
Memmert™ Entry Port 57 mm for UF160plus > For Use With: UF160plus
Spot an opportunity for improvement?Share a Content Correction