Learn More
Memmert™ Entry Port 57 mm for UF750plus
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF750plusF8
Product Code. 17589605
  • 3460.00 SEK / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Description
  • Entry port with 57 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
Specifications
UF750plus Oven
Entry Port
57 mm
Product Title
Memmert™ Entry Port 57 mm for UF750plus > For Use With: UF750plus