Learn More
Lauda™ Tubing
Created from EPDM rubber. Lauda™ EPDM Polymer Tubing is excellent for use with all bath liquids except for Ultra 350 and mineral oils.
Brand:  Lauda™ RKJ 111
Additional Details : Weight : 30.00000kg
Product Code. 10406722
  • 190.00 SEK / 1m
Estimated Shipment: 22-11-2024
to see stock.
Description
Description
  • Made from EPDM
Specifications
Specifications
10°C to 90°C
Tubing
9 mm