missing translation for 'onlineSavingsMsg'
Learn More

Epstein Barr Virus Antibody Cocktail, Invitrogen™

Antibody Cocktail

Brand:  Invitrogen MA512172

Product Code. 11384583

  • 4405.00 SEK / 500µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

MA5-12172 targets Epstein-Barr Virus in IHC (P) applications and shows reactivity with Bacteria samples. The MA5-12172 immunogen is eBV-encoded recombinant latent membrane protein.

Epstein-Barr virus (EBV) is a human herpes virus, which is associated with conditions such as Hodgkin's disease and Burkitt's Lymphoma and is the causative agent in mononucleosis in adolescents. EBV latently infects B lymphocytes. Infected B cells express EBV nuclear antigens and latent proteins LMP1, LMP2A and LMP2B. LMP2A forms aggregates in the plasma membranes of B lymphocytes, where it functions as a negative regulator of the Src and Syk protein tyrosine kinases. Studies show that LMP2A blocks B-cell receptor (BCR) signal transduction in EBV immortalized B cells in vitro and may play an important role in maintaining a latent EBV infection within the peripheral blood B cells of infected individuals.EBNA-1 is the only viral protein consistently expressed in all malignancies associated with EBV.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Epstein Barr Virus
Cocktail
Conc. Not Determined
tissue culture supernatant with 0.09% sodium azide
Mouse
500 μL
Primary
4° C
Liquid
Immunohistochemistry (Paraffin)
CS1, CS2, CS3, CS4, CS2, CS3, CS4
Unconjugated
EBV
EBV-encoded recombinant latent membrane protein
RUO
Virus
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.