missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2 (HER-2) Monoclonal Antibody (6C2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515702
This item is not returnable.
View return policy
Description
MA5-15702 targets ERBB2 in indirect ELISA, FACS, IHC, and WB applications and shows reactivity with Human samples. The MA5-15702 immunogen is purified recombinant fragment of human ERBB2(aa750-987) expressed in E. Coli. MA5-15702 detects ERBB2 which has a predicted molecular weight of approximately 180kDa.
ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.Specifications
ErbB2 (HER-2) | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P04626 | |
ERBB2 | |
Purified recombinant fragment of human ERBB2(aa750-987) expressed in E. Coli. | |
RUO | |
2064 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
6C2 | |
Unconjugated | |
ERBB2 | |
avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb-b2 avian erythroblastic leukemia viral oncoprotein 2; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian) | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction